Learn More
Biomatik Corporation Human Angiopoietin-1 (Angpt1) Elisa Kit
Supplier:  Biomatik Corporation EKC31139
Encompass_Preferred
Human Angiopoietin-1 (Angpt1) Elisa Kit for detecting Angiopoietin 1 in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.091ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15389. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3569