Learn More
Biomatik Corporation Human Eotaxin 2/Ccl24 Elisa Kit
Supplier:  Biomatik Corporation EKC31214
Encompass_Preferred
Human Eotaxin 2/Ccl24 Elisa Kit for detecting Chemokine (C-C Motif) Ligand 24 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 67.660pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O00175; CCL24. Target Synonyms: C C motif chemokine 24; C-C motif chemokine 24; CCL24; CCL24_HUMAN; Chemokine CC Motif Ligand 24; CK beta 6; CK-beta-6; Ckb6; Eosinophil chemotactic protein 2; Eotaxin-2; MPIF 2; MPIF-2; MPIF2; Myeloid progenitor inhibitory factor 2; SCYA24; Small inducible cytokine A24; Small inducible cytokine subfamily A (Cys-Cys) member 24; Small-inducible cytokine A24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3576