Learn More
Biomatik Corporation Human Hemoglobin (Hb) Elisa Kit
Supplier:  Biomatik Corporation EKC31233
Encompass_Preferred
Human Hemoglobin (Hb) Elisa Kit for detecting Hemoglobin (Hb) in serum, plasma, lysate for rbc. Species Reactivity: Human (Homo sapiens). Detection Range: 0.244-1000ng/ml. Sensitivity: 0.244ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3578