Learn More
Biomatik Corporation Human Macrophage Stimulating Protein, Msp Elisa Kit
Supplier:  Biomatik Corporation EKC31277
Encompass_Preferred
Human Macrophage Stimulating Protein, Msp Elisa Kit for detecting Macrophage Stimulating 1 (Hepatocyte Growth Factor-Like) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P26927; MST1. Target Synonyms: D3F15S2; DNF15S2; Hepatocyte growth factor like protein alpha chain; Hepatocyte growth factor like protein; Hepatocyte growth factor like protein beta chain; Hepatocyte growth factor like protein homolog; Hepatocyte growth factor-like protein beta chain; HGFL; HGFL_HUMAN; Macrophage stimulating 1 (hepatocyte growth factor like); Macrophage stimulatory protein; Macrophage-stimulating protein; MSP; MST1; NF15S2; OTTHUMP00000208927. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3580