Learn More
Biomatik Corporation Human Tissue Factor Pathway Inhibitor, Tfpi Elisa Kit
Supplier:  Biomatik Corporation EKC31335
Encompass_Preferred
Human Tissue Factor Pathway Inhibitor, Tfpi Elisa Kit for detecting Tissue Factor Pathway Inhibitor (Lipoprotein-Associated Coagulation Inhibitor) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.782-50ng/ml. Sensitivity: 0.293ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10646; TFPI. Target Synonyms: Anti convertin; EPI; Extrinsic pathway inhibitor; LACI; Lipoprotein associated coagulation inhibitor; Lipoprotein-associated coagulation inhibitor; TFI; TFPI 1; TFPI; TFPI1; TFPI1_HUMAN; Tissue factor pathway inhibitor (lipoprotein associated coagulation inhibitor); Tissue factor pathway inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3587