Learn More
Biomatik Corporation Human Tyrosine-Protein Kinase Receptor Ufo (Axl) Elisa Kit
Supplier:  Biomatik Corporation EKC31350
Encompass_Preferred
Human Tyrosine-Protein Kinase Receptor Ufo (Axl) Elisa Kit for detecting Tyrosine-Protein Kinase Receptor Ufo (Axl) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P30530. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3591