Learn More
Biomatik Corporation Human Visceral Adipose-Specific Serine Protease Inhibitor, Vaspin Elisa Kit
Supplier:  Biomatik Corporation EKC31356
Encompass_Preferred
Human Visceral Adipose-Specific Serine Protease Inhibitor, Vaspin Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade A (Alpha-1 Antiproteinase, Antitrypsin), Member 12 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8IW75; SERPINA12. Target Synonyms: ENSG00000165953; OL 64; OL-64; OL64; Serine (or cysteine) proteinase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 12; Serpin A12; Serpin A12 precursor; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 12; Serpina12; SPA12_HUMAN; Vaspin; Visceral adipose specific serpin; Visceral adipose tissue derived serine protease inhibitor; Visceral adipose tissue-derived serine protease inhibitor; Visceral adipose-specific serpin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3593