Learn More
Biomatik Corporation Mouse Apolipoprotein A5 (Apo-A5) Elisa Kit
Supplier:  Biomatik Corporation EKC31380
Encompass_Preferred
Mouse Apolipoprotein A5 (Apo-A5) Elisa Kit for detecting Apolipoprotein A-V in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q8C7G5; Apoa5. Target Synonyms: Apoa5; Rap3Apolipoprotein A-V; Apo-AV; ApoA-V; Apolipoprotein A5; Regeneration-associated protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3595