Learn More
Biomatik Corporation Mouse Macrophage-Derived Chemokine, Mdc Elisa Kit
Supplier:  Biomatik Corporation EKC31412
Encompass_Preferred
Mouse Macrophage-Derived Chemokine, Mdc Elisa Kit for detecting Chemokine (C-C Motif) Ligand 22 in serum, plasma, tissue homogenates, cell culture supernates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 23.672pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O88430; Ccl22. Target Synonyms: Ccl22; Abcd1; Scya22C-C motif chemokine 22; Activated B and dendritic cell-derived; CC chemokine ABCD-1; Small-inducible cytokine A22. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3602