Learn More
Biomatik Corporation Pig apolipoprotein E (Apo-E) ELISA Kit
Supplier:  Biomatik Corporation EKC31444
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 31.25 ng/mL-2000 ng/mL. Sensitivity: 7.81 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P18650. Target Synonyms: AD2, LDLCQ5, LPG, MGC1571, apolipoprotein E3. Restriction: For Research Use Only.
Catalog No. 50-229-3612