Learn More
Biomatik Corporation Rat L-Selectin ELISA kit
Supplier:  Biomatik Corporation EKC31494
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 78.125 pg/mL-5000 pg/mL. Sensitivity: 64 pg/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P30836. Target Synonyms: CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1, CD62 antigen-like family member L|gp90-MEL|leukocyte surface antigen Leu-8|leukocyte-endothelial cell adhesion molecule 1|lymph node homing. Restriction: For Research Use Only.
Catalog No. 50-229-3623