Learn More
Biomatik Corporation Sheep Albumin (Alb) Elisa Kit
Supplier:  Biomatik Corporation EKC31504
Encompass_Preferred
Sheep Albumin (Alb) Elisa Kit for detecting Albumin in serum, plasma, cell culture supernates, urine, tissue homogenates. Species Reactivity: Sheep (Ovis aries). Detection Range: 350-21.88ug/ml. Sensitivity: 5.5ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P14639; ALB. Target Synonyms: ALBSerum albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3625