Learn More
Biomatik Corporation Bovine Alpha-2-Macroglobulin (A2M) Elisa Kit
Supplier:  Biomatik Corporation EKC31545
Encompass_Preferred
Bovine Alpha-2-Macroglobulin (A2M) Elisa Kit for detecting Alpha-2-Macroglobulin in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ug/ml. Sensitivity: 0.078ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q7SIH1; A2M. Target Synonyms: A2MAlpha-2-macroglobulin; Alpha-2-M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3627