Learn More
Biomatik Corporation Bovine Beta-Hydroxybutyric Acid,β-OHB ELISA Kit
Supplier:  Biomatik Corporation EKC31562
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Bos taurus (Bovine). Detection Range: 50 nmol/mL-800 nmol/mL. Sensitivity: 9.5 nmol/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: beta-Hydroxybutyric acid, beta-OHB. Restriction: For Research Use Only.
Catalog No. 50-229-3631