Learn More
Biomatik Corporation Human beta 2 transferrin (β2TF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32844
Encompass_Preferred
The Human beta 2 transferrin (β2TF) ELISA Kit allows for quantitative determination of beta 2 transferrin (β2TF) in serum, cerebrospinal fluid. Species Reactivity: Human. Detection Range: 1.25 μg/mL-80 μg/mL. Sensitivity: 0.31μg/mL. Format: 96-Well Plate. Assay Type: ELISA. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7715