Learn More
Biomatik Corporation Bovine Heat Shock Protein 70, Hsp-70 Elisa Kit
Supplier:  Biomatik Corporation EKC31646
Encompass_Preferred
Bovine Heat Shock Protein 70, Hsp-70 Elisa Kit for detecting Heat Shock Protein 70, Hsp-70 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.25-20ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q27975 ; HSPA1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3639