Learn More
Biomatik Corporation Bovine Lipopolysaccharides (Lps) Elisa Kit
Supplier:  Biomatik Corporation EKC31717
Encompass_Preferred
Bovine Lipopolysaccharides (Lps) Elisa Kit for detecting Lipopolysaccharides (Lps) in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3646