Learn More
Biomatik Corporation Bovine Myoglobin (Mb) Elisa Kit
Supplier:  Biomatik Corporation EKC31740
Encompass_Preferred
Bovine Myoglobin (Mb) Elisa Kit for detecting Myoglobin in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02192; MB. Target Synonyms: MB; Myoglobin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3648