Learn More
Biomatik Corporation Bovine Pentraxin-Related Protein Ptx3 (Ptx3) Elisa Kit
Supplier:  Biomatik Corporation EKC31752
Encompass_Preferred
Bovine Pentraxin-Related Protein Ptx3 (Ptx3) Elisa Kit for detecting Pentraxin-Related Gene, Rapidly Induced By Il-1 Beta in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 2.344-150ng/ml. Sensitivity: 0.586ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q0VCG9; PTX3. Target Synonyms: PTX3; Pentraxin-related protein PTX3; Pentaxin-related protein PTX3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3651