Learn More
Biomatik Corporation Bovine Ubiquitin-Like Protein Isg15 (Isg15) Elisa Kit
Supplier:  Biomatik Corporation EKC31821
Encompass_Preferred
Bovine Ubiquitin-Like Protein Isg15 (Isg15) Elisa Kit for detecting Isg15 Ubiquitin-Like Modifier in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O02741; ISG15. Target Synonyms: ISG15; G1P2; ISG17; UCRP; Ubiquitin-like protein ISG15; Interferon-stimulated gene product 17; Ubiquitin cross-reactive protein; BoUCRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3658