Learn More
Biomatik Corporation Bovine Vitamin D-Binding Protein (Gc) Elisa Kit
Supplier:  Biomatik Corporation EKC31824
Encompass_Preferred
Bovine Vitamin D-Binding Protein (Gc) Elisa Kit for detecting Group-Specific Component (Vitamin D Binding Protein) in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ug/ml. Sensitivity: 0.039ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q3MHN5; GC. Target Synonyms: GCVitamin D-binding protein; DBP; VDB; Gc-globulin; Group-specific component. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3660