Learn More
Biomatik Corporation Canine Free Thyroxine, Ft4 Elisa Kit
Supplier:  Biomatik Corporation EKC31851
Encompass_Preferred
Canine Free Thyroxine, Ft4 Elisa Kit for detecting Free Thyroxine, Ft4 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 7.5-120ng/ml. Sensitivity: 7.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3667