Learn More
Biomatik Corporation Canine Luteinizing Hormone, Lh Elisa Kit
Supplier:  Biomatik Corporation EKC31863
Encompass_Preferred
Canine Luteinizing Hormone, Lh Elisa Kit for detecting Luteinizing Hormone, Lh in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.75-60mIU/ml. Sensitivity: 0.94mIU/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P18842; LHB. Target Synonyms: LHBLutropin subunit beta; Luteinizing hormone subunit beta; LH-B; LSH-B; LSH-beta; Lutropin beta chain; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3669