Learn More
Biomatik Corporation Canine Thyroid Stimulating Hormone, Tsh Elisa Kit
Supplier:  Biomatik Corporation EKC31886
Encompass_Preferred
Canine Thyroid Stimulating Hormone, Tsh Elisa Kit for detecting Thyroid Stimulating Hormone, Tsh in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.625-10uIU/ml. Sensitivity: 0.625uIU/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P54828; TSHB. Target Synonyms: TSHB; Thyrotropin subunit beta; Thyroid-stimulating hormone subunit beta; TSH-B; TSH-beta; Thyrotropin beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3677