Learn More
Biomatik Corporation Chicken Cluster Of Differentiation 4 (Cd4) Elisa Kit
Supplier:  Biomatik Corporation EKC31913
Encompass_Preferred
Chicken Cluster Of Differentiation 4 (Cd4) Elisa Kit for detecting Cluster Of Differentiation 4(Cd4) in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 3.125-800ng/ml. Sensitivity: 3.125ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q9W6V7; CD4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3679