Learn More
Biomatik Corporation Chicken secretory immunoglobulin A,SIgA ELISA Kit
Supplier:  Biomatik Corporation EKC31987
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Gallus gallus (Chicken). Detection Range: 0.234 μg/mL-60 μg/mL. Sensitivity: 0.058 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3690