Learn More
Biomatik Corporation Dog Insulin-Like Growth Factor Binding Protein 3 (Igfbp3) Elisa Kit
Supplier:  Biomatik Corporation EKC32060
Encompass_Preferred
Dog Insulin-Like Growth Factor Binding Protein 3 (Igfbp3) Elisa Kit for detecting Insulin-Like Growth Factor Binding Protein 3 (Igfbp3) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: F1PQ91. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3699