Learn More
Biomatik Corporation Dog Motilin (Mln) Elisa Kit
Supplier:  Biomatik Corporation EKC32092
Encompass_Preferred
Dog Motilin (Mln) Elisa Kit for detecting Motilin in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19863; MLN. Target Synonyms: MLN; Motilin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3703