Learn More
Biomatik Corporation Dog Vascular Endothelial Cell Growth Factor, Vegf Elisa Kit
Supplier:  Biomatik Corporation EKC32129
Encompass_Preferred
Dog Vascular Endothelial Cell Growth Factor, Vegf Elisa Kit for detecting Vascular Endothelial Growth Factor A in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9MYV3; VEGFA. Target Synonyms: VEGFA; VEGF; Vascular endothelial growth factor A; VEGF-A; Vascular permeability factor; VPF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3709