Learn More
Biomatik Corporation Fish Cytochrome P450 1A1 (Cyp1A1) Elisa Kit
Supplier:  Biomatik Corporation EKC32148
Encompass_Preferred
Fish Cytochrome P450 1A1 (Cyp1A1) Elisa Kit for detecting Cytochrome P450, Family 1, Subfamily A, Polypeptide 1 in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 75-1200pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q6JZS3; cyp1a1. Target Synonyms: cyp1a1; cyp1aCytochrome P450 1A1; EC 1.14.14.1; CYPIA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3714