Learn More
Biomatik Corporation Fish Thyroid Stimulating Hormone (Tsh) Elisa Kit
Supplier:  Biomatik Corporation EKC32168
Encompass_Preferred
Fish Thyroid Stimulating Hormone (Tsh) Elisa Kit for detecting Thyroid Stimulating Hormone(Tsh) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 2.5-40uIU/ml. Sensitivity: 2.5uIU/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P37240; tshb. Target Synonyms: tshb; Thyrotropin subunit beta; Thyroid-stimulating hormone subunit beta; TSH-B; TSH-beta; Thyrotropin beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3716