Learn More
Biomatik Corporation Goat Adiponectin (Adp) Elisa Kit
Supplier:  Biomatik Corporation EKC32173
Encompass_Preferred
Goat Adiponectin (Adp) Elisa Kit for detecting Adiponectin(Adp) in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: U6A087. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3718