Learn More
Biomatik Corporation Goat Interleukin 12, Il-12/P40 Elisa Kit
Supplier:  Biomatik Corporation EKC32208
Encompass_Preferred
Goat Interleukin 12, Il-12/P40 Elisa Kit for detecting Interleukin 12B (Natural Killer Cell Stimulatory Factor 2, Cytotoxic Lymphocyte Maturation Factor 2, P40) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 9.38-600pg/ml. Sensitivity: 2.34pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P68221; IL12B. Target Synonyms: IL12B; Interleukin-12 subunit beta; IL-12B; Cytotoxic lymphocyte maturation factor 40 kDa subunit; CLMF p40; IL-12 subunit p40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3724