Learn More
Biomatik Corporation Hamster Putative Phospholipase B-Like 2 (Plbd2) Elisa Kit
Supplier:  Biomatik Corporation EKC32298
Encompass_Preferred
Hamster Putative Phospholipase B-Like 2 (Plbd2) Elisa Kit for detecting Phospholipase B Domain Containing 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Golden hamster. Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: P76; LAMA-like protein 2; PLB homolog 2; lamina ancestor homolog 2; mannose-6-phosphate protein associated protein p76; phospholipase B-like 2 32 kDa form; phospholipase B-like 2 45 kDa form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3734