Learn More
Biomatik Corporation Horse Prolactin(PRL) ELISA kit
Supplier:  Biomatik Corporation EKC32366
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Equus caballus (Horse). Detection Range: 2.5 ng/mL-1000 ng/mL. Sensitivity: 2.5 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P12420. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3745