Learn More
Biomatik Corporation Human 1, 25-Dihydroxyvitamin D (3) 24-Hydroxylase, Mitochondrial (Cyp24A1) Elisa Kit
Supplier:  Biomatik Corporation EKC32378
Encompass_Preferred
Human 1, 25-Dihydroxyvitamin D (3) 24-Hydroxylase, Mitochondrial (Cyp24A1) Elisa Kit for detecting 1, 25-Dihydroxyvitamin D(3) 24-Hydroxylase, Mitochondrial (Cyp24A1),Cytochrome P450, Family 24, Subfamily A, Polypeptide 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q07973; CYP24A1. Target Synonyms: 1 25 @dihydroxyvitamin D3 24 hydroxylase; 1 25 dihydroxyvitamin D (3) 24 hydroxylase; 1; 1; 25-dihydroxyvitamin D (3) 24-hydroxylase; mitochondrial; 24 OHase; 24-OHase; 25-dihydroxyvitamin D (3) 24-hydroxylase; CP 24; CP24; CP24A_HUMAN; CYP 24; CYP24; CYP24A1; Cytochrome P450 24A1; Cytochrome P450 24A1 mitochondrial; cytochrome P450 CC24; Cytochrome P450 family 24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3747