Learn More
Biomatik Corporation Human 14-3-3 Protein Theta (Ywhaq) Elisa Kit
Supplier:  Biomatik Corporation EKC32385
Encompass_Preferred
Human 14-3-3 Protein Theta (Ywhaq) Elisa Kit for detecting Tyrosine 3-Monooxygenase/Tryptophan 5-Monooxygenase Activation Protein, Theta Polypeptide in serum, plasma, tissue homogenates, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.47-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P27348; YWHAQ. Target Synonyms: 14-3-3 protein T cell; 14-3-3 protein T-cell; 14-3-3 protein tau; 14-3-3 protein theta; 1433T_HUMAN; IC5; Protein HS1; Protein tau; YWHAQ tyrosine 3 monooxygenase/tryptophan 5-monooxygenase activation protein; theta polypeptide; Ywhaq. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3751