Learn More
Biomatik Corporation Human 26S Proteasome Non-Atpase Regulatory Subunit 4 (Psmd4) Elisa Kit
Supplier:  Biomatik Corporation EKC32395
Encompass_Preferred
Human 26S Proteasome Non-Atpase Regulatory Subunit 4 (Psmd4) Elisa Kit for detecting Proteasome (Prosome, Macropain) 26S Subunit, Non-Atpase, 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 34.4-2200pg/ml. Sensitivity: 8.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55036; PSMD4. Target Synonyms: 26S protease subunit S5a; 26S proteasome non ATPase regulatory subunit 4; 26S proteasome non-ATPase regulatory subunit 4; 26S proteasome regulatory subunit rpn10; 26S proteasome regulatory subunit S5A; AF 1; AF; AF1; Angiocidin; Antisecretory factor 1; ASF; DS5a; MCB 1; MCB1; Multiubiquitin chain binding protein; Multiubiquitin chain-binding protein; OTTHUMP00000059963; Prosome macropain; Proteasome (prosome macropain) 26S subunit non ATPase 4; Proteasome 19S S5A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3754