Learn More
Biomatik Corporation Human A Disintegrin And Metalloproteinase With Thrombospondin Motifs 5 (Adamts5) Elisa Kit
Supplier:  Biomatik Corporation EKC32444
Encompass_Preferred
Human A Disintegrin And Metalloproteinase With Thrombospondin Motifs 5 (Adamts5) Elisa Kit for detecting Adam Metallopeptidase With Thrombospondin Type 1 Motif, 5 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UNA0; ADAMTS5. Target Synonyms: A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 5; A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 5; A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif; 5 (aggrecanase 2); A disintegrin-like and metalloprotease with thrombospondin type 1 motif; 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3764