Learn More
Biomatik Corporation Human Acyloxyacyl Hydrolase (Aoah) Elisa Kit
Supplier:  Biomatik Corporation EKC32483
Encompass_Preferred
Human Acyloxyacyl Hydrolase (Aoah) Elisa Kit for detecting Acyloxyacyl Hydrolase (Neutrophil) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P28039; AOAH. Target Synonyms: Acyloxyacyl hydrolase (neutrophil); Acyloxyacyl hydrolase large subunit; AOAH; AOAH_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3770