Learn More
Biomatik Corporation Human Adamts-Like Protein 4 (Adamtsl4) Elisa Kit
Supplier:  Biomatik Corporation EKC32490
Encompass_Preferred
Human Adamts-Like Protein 4 (Adamtsl4) Elisa Kit for detecting Adamts-Like 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UY14; ADAMTSL4. Target Synonyms: ADAMTSL4; TSRC1; PP1396; UNQ2803/PRO34012ADAMTS-like protein 4; ADAMTSL-4; Thrombospondin repeat-containing protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3772