Learn More
Biomatik Corporation Human Adenylyl Cyclase-Associated Protein 1 (Cap1) Elisa Kit
Supplier:  Biomatik Corporation EKC32497
Encompass_Preferred
Human Adenylyl Cyclase-Associated Protein 1 (Cap1) Elisa Kit for detecting Cap, Adenylate Cyclase-Associated Protein 1 (Yeast) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q01518; CAP1. Target Synonyms: Adenylyl cyclase associated protein 1; Adenylyl cyclase-associated protein 1; CAP 1; CAP; CAP; adenylate cyclase associated protein 1 (yeast); CAP1; CAP1 PEN; CAP1-PEN; CAP1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3774