Learn More
Biomatik Corporation Human Adrenomedullin, Adm Elisa Kit
Supplier:  Biomatik Corporation EKC32506
Encompass_Preferred
Human Adrenomedullin, Adm Elisa Kit for detecting Adrenomedullin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20pg/ml. Sensitivity: 0.078pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35318; ADM. Target Synonyms: Adm; ADM precursor; ADML_HUMAN; Adrenomedullin; AM; PAMP; Preproadrenomedullin; Proadrenomedullin N-20 terminal peptide; Proadrenomedullin N20 terminal peptide; ProAM N terminal 20 peptide; ProAM N-terminal 20 peptide; ProAM N20; ProAM-N20; ProAMN20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3778