Learn More
Biomatik Corporation Human A-Kinase Anchor Protein 4 (Akap4) Elisa Kit
Supplier:  Biomatik Corporation EKC32515
Encompass_Preferred
Human A-Kinase Anchor Protein 4 (Akap4) Elisa Kit for detecting A Kinase (Prka) Anchor Protein 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5JQC9; AKAP4. Target Synonyms: A kinase (PRKA) anchor protein 4; A kinase anchor protein 4; A kinase anchor protein 82 kDa; A-kinase anchor protein 4; A-kinase anchor protein 82 kDa; AKAP 4; AKAP 82; AKAP-4; AKAP4; AKAP4_HUMAN; AKAP82; FSC 1; FSC1; hAKAP 82; hAKAP82; HI; Major sperm fibrous sheath protein; p82; PRKA 4; PRKA4; Protein kinase A anchoring protein 4; Protein kinase A-anchoring protein 4; Testis specific gene HI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3780