Learn More
Biomatik Corporation Human Alpha-Crystallin B Chain (Cryab) Elisa Kit
Supplier:  Biomatik Corporation EKC32544
Encompass_Preferred
Human Alpha-Crystallin B Chain (Cryab) Elisa Kit for detecting Crystallin, Alpha B in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02511; CRYAB. Target Synonyms: AACRYA; Alpha B crystallin; Alpha crystallin B chain; Alpha (B)-crystallin; Alpha-crystallin B chain; CRYA2; Cryab; CRYAB_HUMAN; Crystallin alpha B; Crystallin alpha polypeptide 2; CTPP2; Heat shock 20 kD like protein; Heat shock protein beta 5; Heat shock protein beta-5; HspB5; Renal carcinoma antigen NY REN 27; Renal carcinoma antigen NY-REN-27; Rosenthal fiber component. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3787