Learn More
Biomatik Corporation Human Angiotensin Ⅱ Receptor 2 antibody,AT2R-Ab ELISA Kit
Supplier:  Biomatik Corporation EKC32591
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312 U/mL-20 U/mL. Sensitivity: 0.078 U/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: angiotensin 2 receptor 2 antibody, AT2R-Ab. Restriction: For Research Use Only.
Catalog No. 50-229-3794