Learn More
Biomatik Corporation Human Anosmin-1 (Kal1) Elisa Kit
Supplier:  Biomatik Corporation EKC32608
Encompass_Preferred
Human Anosmin-1 (Kal1) Elisa Kit for detecting Kallmann Syndrome 1 Sequence in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P23352; ANOS1. Target Synonyms: Adhesion molecule-like X-linked; ADMLX; Anosmin-1; HHA; KAL; KAL1; KALIG 1; KALIG1; Kallmann syndrome 1 sequence (anosmin 1); Kallmann syndrome 1 sequence; Kallmann syndrome interval gene 1; Kallmann syndrome protein; KALM; KALM_HUMAN; KMS; OTTHUMP00000022873. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3797