Learn More
Biomatik Corporation Human Apolipoprotein F (Apof) Elisa Kit
Supplier:  Biomatik Corporation EKC32763
Encompass_Preferred
Human Apolipoprotein F (Apof) Elisa Kit for detecting Apolipoprotein F in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 9.38-600ng/ml. Sensitivity: 2.34ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q13790; APOF. Target Synonyms: Apo-F; Apof; APOF_HUMAN; Apolipoprotein F; DKFZp781G18150; Lipid transfer inhibitor protein; LTIP; MGC22520. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3806