Learn More
Biomatik Corporation Human B- And T-Lymphocyte Attenuator (Btla) Elisa Kit
Supplier:  Biomatik Corporation EKC32819
Encompass_Preferred
Human B- And T-Lymphocyte Attenuator (Btla) Elisa Kit for detecting B And T Lymphocyte Associated in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.45-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q7Z6A9; BTLA. Target Synonyms: B and T lymphocyte associated protein; B and T lymphocyte attenuator; B and T lymphocyte associated; B- and T-lymphocyte attenuator; B- and T-lymphocyte-associated protein; BTLA; BTLA_HUMAN; BTLA1; CD272; CD272 antigen; FLJ16065; MGC129743. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3814