Learn More
Biomatik Corporation Human Camp Response Element-Binding Protein (Creb) Elisa Kit
Supplier:  Biomatik Corporation EKC32964
Encompass_Preferred
Human Camp Response Element-Binding Protein (Creb) Elisa Kit for detecting Camp Responsive Element Binding Protein 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16220; CREB1. Target Synonyms: Active transcription factor CREB; cAMP response element binding protein 1; cAMP response element binding protein; cAMP responsive element binding protein 1; cAMP-responsive element-binding protein 1; CREB; CREB-1; CREB1; CREB1_HUMAN; Cyclic AMP-responsive element-binding protein 1; MGC9284; OTTHUMP00000163864; OTTHUMP00000163865; OTTHUMP00000206660; OTTHUMP00000206662; OTTHUMP00000206667; Transactivator protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3838